Merck
  • Three-dimensional genome architecture coordinates key regulators of lineage specification in mammary epithelial cells.
Three-dimensional genome architecture coordinates key regulators of lineage specification in mammary epithelial cells.
Cell genomics (2023-11-29)
Michael J G Milevskiy, Hannah D Coughlan, Serena R Kane, Timothy M Johanson, Somayeh Kordafshari, Wing Fuk Chan, Minhsuang Tsai, Elliot Surgenor, Stephen Wilcox, Rhys S Allan, Yunshun Chen, Geoffrey J Lindeman, Gordon K Smyth, Jane E Visvader
ABSTRACT
Although lineage-specific genes have been identified in the mammary gland, little is known about the contribution of the 3D genome organization to gene regulation in the epithelium. Here, we describe the chromatin landscape of the three major epithelial subsets through integration of long- and short-range chromatin interactions, accessibility, histone modifications, and gene expression. While basal genes display exquisite lineage specificity via distal enhancers, luminal-specific genes show widespread promoter priming in basal cells. Cell specificity in luminal progenitors is largely mediated through extensive chromatin interactions with super-enhancers in gene-body regions in addition to interactions with polycomb silencer elements. Moreover, lineage-specific transcription factors appear to be controlled through cell-specific chromatin interactivity. Finally, chromatin accessibility rather than interactivity emerged as a defining feature of the activation of quiescent basal stem cells. This work provides a comprehensive resource for understanding the role of higher-order chromatin interactions in cell-fate specification and differentiation in the adult mouse mammary gland.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Sigma-Aldrich
Anti-RNA polymerase II Antibody, clone CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross adsorbed antibody produced in guinea pig, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross adsorbed antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross adsorbed antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit