Merck
  • The metabolic function of pyruvate kinase M2 regulates reactive oxygen species production and microbial killing by neutrophils.
The metabolic function of pyruvate kinase M2 regulates reactive oxygen species production and microbial killing by neutrophils.
Nature communications (2023-07-18)
Juliana Escher Toller-Kawahisa, Carlos Hiroji Hiroki, Camila Meirelles de Souza Silva, Daniele Carvalho Nascimento, Gabriel Azevedo Públio, Timna Varela Martins, Luis Eduardo Alves Damasceno, Flávio Protásio Veras, Paula Ramos Viacava, Fábio Yuji Sukesada, Emily Anne Day, Alessia Zotta, Tristram Alexander Jasper Ryan, Rodrigo Moreira da Silva, Thiago Mattar Cunha, Norberto Peporine Lopes, Fernando de Queiroz Cunha, Luke Anthony John O'Neill, José Carlos Alves-Filho
ABSTRACT
Neutrophils rely predominantly on glycolytic metabolism for their biological functions, including reactive oxygen species (ROS) production. Although pyruvate kinase M2 (PKM2) is a glycolytic enzyme known to be involved in metabolic reprogramming and gene transcription in many immune cell types, its role in neutrophils remains poorly understood. Here, we report that PKM2 regulates ROS production and microbial killing by neutrophils. Zymosan-activated neutrophils showed increased cytoplasmic expression of PKM2. Pharmacological inhibition or genetic deficiency of PKM2 in neutrophils reduced ROS production and Staphylococcus aureus killing in vitro. In addition, this also resulted in phosphoenolpyruvate (PEP) accumulation and decreased dihydroxyacetone phosphate (DHAP) production, which is required for de novo synthesis of diacylglycerol (DAG) from glycolysis. In vivo, PKM2 deficiency in myeloid cells impaired the control of infection with Staphylococcus aureus. Our results fill the gap in the current knowledge of the importance of lower glycolysis for ROS production in neutrophils, highlighting the role of PKM2 in regulating the DHAP and DAG synthesis to promote ROS production in neutrophils.
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Zymosan A from Saccharomyces cerevisiae, for inducing inflamatory response
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
PFKFB3 Inhibitor, 3PO, PFKFB3 Inhibitor, 3PO, is a cell-permeable, selective inhibitor of PFK-2 (6-phosphofructo-2-Kinase) activity.