Merck
HDGFRP3 interaction with 53BP1 promotes DNA double-strand break repair.
Nucleic acids research (2023-02-17)
Zhen Zhang, William E Samsa, Yanyan De, Fan Zhang, Ofer Reizes, Alexandru Almasan, Zihua Gong
ABSTRACT
The 53BP1-dependent end-joining pathway plays a critical role in double-strand break (DSB) repair. However, the regulators of 53BP1 in chromatin remain incompletely characterized. In this study, we identified HDGFRP3 (hepatoma-derived growth factor related protein 3) as a 53BP1-interacting protein. The HDGFRP3-53BP1 interaction is mediated by the PWWP domain of HDGFRP3 and the Tudor domain of 53BP1. Importantly, we observed that the HDGFRP3-53BP1 complex co-localizes with 53BP1 or γH2AX at sites of DSB and participates in the response to DNA damage repair. Loss of HDGFRP3 impairs classical non-homologous end-joining repair (NHEJ), curtails the accumulation of 53BP1 at DSB sites, and enhances DNA end-resection. Moreover, the HDGFRP3-53BP1 interaction is required for cNHEJ repair, 53BP1 recruitment at DSB sites, and inhibition of DNA end resection. In addition, loss of HDGFRP3 renders BRCA1-deficient cells resistant to PARP inhibitors by facilitating end-resection in BRCA1 deficient cells. We also found that the interaction of HDGFRP3 with methylated H4K20 was dramatically decreased; in contrast, the 53BP1-methylated H4K20 interaction was increased after ionizing radiation, which is likely regulated by protein phosphorylation and dephosphorylation. Taken together, our data reveal a dynamic 53BP1-methylated H4K20-HDGFRP3 complex that regulates 53BP1 recruitment at DSB sites, providing new insights into our understanding of the regulation of 53BP1-mediated DNA repair pathway.
Sigma-Aldrich
Anti-RPA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, culture supernatant, clone A3S, Upstate®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)