Merck
  • DOT1L regulates MTDH-mediated angiogenesis in triple-negative breast cancer: intermediacy of NF-κB-HIF1α axis.
DOT1L regulates MTDH-mediated angiogenesis in triple-negative breast cancer: intermediacy of NF-κB-HIF1α axis.
The FEBS journal (2022-08-27)
Praveen Kumar Neeli, Shashikanta Sahoo, Santosh Karnewar, Gajalakshmi Singuru, Sriravali Pulipaka, Sandhya Annamaneni, Srigiridhar Kotamraju
ABSTRACT
DOT1L, a specific H3K79 methyltransferase, has a tumour-promoting role in various cancers, including triple-negative breast cancer (TNBC). However, the molecular mechanism by which the deregulated DOT1L promotes cancer progression is unclear. Herein, we show that a significantly higher basal level of DOTL1 strongly correlates with MTDH, an oncogene, in clinical TNBC patient cohorts and mediates TNBC progression by enhancing MTDH-induced angiogenesis. In parallel, severe combined immunodeficiency mice-bearing MDA-MB-231 cells with MTDH-Wt or MTDHΔ7 (spliced isoform of MTDH) overexpression constructs showed enhanced blood vessel formations at the tumour site in comparison with control groups. Selective inhibition of DOT1L by EPZ004777, a specific DOT1L inhibitor, or siDOT1L, significantly impaired MTDH-induced proliferation, invasion and angiogenic markers expression in TNBC cells. ChIP assay revealed that Dot1L promotes MTDH-Wt/Δ7 transcription by increasing H3K79me3 levels on its promoter. Dot1L depletion reversed this effect. Mechanistically, DOT1L-induced MTDH caused enhanced nuclear factor kappa B (NF-κB) occupancy on the hypoxia-inducible factor1α (HIF1α) promoter and increased its transcription, leading to elevated levels of proangiogenic mediators in TNBC cells. Moreover, the condition media obtained from MDA-MB-231 cells stably expressing either MTDH-Wt or MTDHΔ7 treated with EPZ004777 or Bay-11-7082 (NF-κB inhibitor) or FM19G11 (HIF1α inhibitor) significantly inhibited MTDH-induced tube formation in human umbilical vein endothelial cells, rat aortic ring sprouting and vessel formations by chick chorioallantoic membrane assay mimicking physiological angiogenic vasculature. Collectively, our findings reveal a novel epigenetic regulation of MTDH by DOTL1, which drives angiogenesis, and that the therapeutic disruption of the DOT1L-MTDH-NF-κB-HIF1α axis may have usefulness in the management of TNBC.
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-VEGFR2 (Ab-1059) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MTDH antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture