Merck
  • Microtubule-targeting agents impair kinesin-2-dependent nuclear transport of β-catenin: Evidence of inhibition of Wnt/β-catenin signaling as an important antitumor mechanism of microtubule-targeting agents.
Microtubule-targeting agents impair kinesin-2-dependent nuclear transport of β-catenin: Evidence of inhibition of Wnt/β-catenin signaling as an important antitumor mechanism of microtubule-targeting agents.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2021-03-21)
Anuradha Kumari, Omprakash Shriwas, Shailendra Sisodiya, Manas K Santra, Sankar K Guchhait, Rupesh Dash, Dulal Panda
ABSTRACT
An aberrant accumulation of nuclear β-catenin is closely associated with the augmentation of cancer malignancy. In this work, we report that several microtubule-targeting agents (MTAs) such as vinblastine, taxol, and C12 (combretastatin-2-aminoimidazole analog) inhibit Wnt/β-catenin signaling in oral squamous cell carcinoma (OSCC). We showed that the inhibition of microtubule dynamics by MTAs decreased the level of β-catenin by increasing Axin and adenomatous polyposis coli levels and reducing the level of dishevelled. Furthermore, MTAs strongly reduced the localization of β-catenin in the nucleus. The reduction in the level of nuclear β-catenin was neither due to the degradation of β-catenin in the nucleus nor due to an increase in the export of nuclear β-catenin from the nucleus. A motor protein kinesin-2 was found to assist the nuclear transportation of β-catenin. Interestingly, Wnt/β-catenin signaling antagonist treatment synergized with MTAs and the activators of Wnt/β-catenin signaling antagonized with the MTAs. C12 potently suppressed the growth of 4-Nitroquinoline 1-oxide-induced OSCC in the tongue of C57 black 6 mice and also abrogated Wnt/β-catenin signaling pathway in the tumor. Our results provide evidence that the decrease in Wnt/β-catenin signaling is an important antitumor effect of MTAs and the combined use of MTAs with Wnt/β-catenin signaling antagonists could be a promising strategy for cancer chemotherapy.
Sigma-Aldrich
Anti-DVL1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution, antigen mol wt ~85 kDa
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-APC Antibody, clone CC-1, clone CC-1, from mouse
Sigma-Aldrich
Anti-Axin-1 Antibody, from rabbit, purified by affinity chromatography