Merck
  • Development of bead based multiplexed immunoassay for evaluation of midkine, syndecan-1, and ANGPTL4 in patient serum.
Development of bead based multiplexed immunoassay for evaluation of midkine, syndecan-1, and ANGPTL4 in patient serum.
Journal of immunoassay & immunochemistry (2018-01-09)
Imad Tarhoni, Cristina L Fhied, Mark Pool, Michael J Liptay, Philip Bonomi, Christopher W Seder, Jeffrey A Borgia
ABSTRACT
Angiogenesis is associated with tumor progression in a range of malignancies. Herein, we develop custom immunobead assays for several mechanistically important targets and evaluated these against sera from cohorts of non-small cell lung cancer (NSCLC) patients. Antigen "capture" antibodies for midkine, syndecan-1, and ANGPTL4 were independently conjugated to MagPlex® Microspheres using standard carbodiimide/NHS-based chemistry. These reagents served as the basis for quantitative sandwich assay assembly using biotinylated detection antibodies and R-phycoerythrin-conjugated streptavidin reporter system. Standard curves were created using dilution series of recombinant target proteins with assay performance characteristics calculated, accordingly. Finally, we evaluated a range of serum samples from NSCLC patients (n = 32) to verify assay performance. Multiplexed assays for midkine, syndecan-1, and ANGPTL4 were developed with three orders of magnitude in dynamic range, excellent intra- and inter-assay precision, and accuracy parameters (<10%, and <15% variability, respectively). Detection and quantifications limits were suitable for the three assays to efficiently evaluate sera across a range of disease stages with a four-fold dilution factor. We successfully developed and analytically validated a 3-plex immunobead assay for quantifying midkine, syndecan-1, and ANGPTL4 in patient sera. This multiplexed assay will provide an important tool for future studies delineating the role of angiogenesis in lung cancer progression.
Product Description
Millipore
MILLIPLEX® Human Liver Protein Magnetic Bead Panel - Metabolism Multiplex Assay, The Human Liver Protein Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 9 liver protein biomarkers in human serum, plasma and tissue/cell culture samples.
Millipore
MILLIPLEX® Human Circulating Cancer Biomarker Magnetic Bead Panel 4 - Cancer Multiplex Assay