Merck
  • Time-lapse imaging of Drosophila testis for monitoring actin dynamics and sperm release.
Time-lapse imaging of Drosophila testis for monitoring actin dynamics and sperm release.
STAR protocols (2022-01-04)
Tushna Kapoor, Pankaj Dubey, Krishanu Ray
ABSTRACT
Here we describe a simple step-by-step protocol for collecting high-resolution, time-lapse images of intact Drosophila testis ex vivo for a limited period using a confocal microscope, with minimum photo-toxic damage, to monitor spermatid individualization, coiling, and release. The F-actin dynamics during spermatid morphogenesis can be further investigated through laser ablations, Fluorescence-Recovery-After-Photobleaching, and drug treatments, using this protocol. For complete details on the use and execution of this protocol, please refer to Dubey et al. (2016), Dubey et al. (2019), and Kapoor et al. (2021).
Sigma-Aldrich
Dispase® I, protease
Sigma-Aldrich
Jasplakinolide, ≥97% (HPLC)
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Formin FH2 Domain Inhibitor, SMIFH2, The Formin FH2 Domain Inhibitor, SMIFH2, also referenced under CAS 340316-62-3, controls the biological activity of Formin FH2 Domain. This small molecule/inhibitor is primarily used for Membrane applications.
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Rho Kinase Inhibitor III, Rockout, The Rho Kinase Inhibitor III, Rockout, also referenced under CAS 7272-84-6, controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.