Merck
  • Rinderpest virus blocks type I and type II interferon action: role of structural and nonstructural proteins.
Rinderpest virus blocks type I and type II interferon action: role of structural and nonstructural proteins.
Journal of virology (2006-07-15)
Sambit K Nanda, Michael D Baron
ABSTRACT
Rinderpest virus (RPV) is a paramyxovirus closely related to the human pathogen Measles virus. It causes severe disease in cattle, buffalo, and some wild animals; although it can infect humans, it does not cause disease. Here, we demonstrate that RPV blocks the action of both type I (alpha) and type II (gamma) interferons (IFNs) by blocking the phosphorylation and nuclear translocation of STAT1 and STAT2 and that this block is not related to species specificity. In addition, both wild-type virulent and vaccine strains of the virus blocked IFN action. Unlike the case with some other paramyxoviruses, neither STAT1 nor STAT2 is degraded upon virus infection. STAT1 is bound by both the viral structural protein P, and thereby recruited to concentrations of viral protein in the cell, and the nonstructural protein V. Although both P and V proteins bind to STAT1 and can block IFN action when expressed in transfected cells, the IFN antagonist activity of the P protein is weaker than that of the V protein. The viral C protein also seems to weakly block IFN-induced activation of STAT1 in transfection experiments. However, studies with knockout viruses showed that the viral V protein appears to be the dominant inhibitor of IFN signaling in the context of virus infection, since prevention of viral V expression restored the IFN sensitivity of infected cells. Although a change in the distribution pattern of STAT2 was observed in virus-infected cells, STAT2 was not bound by any viral protein.
Sigma-Aldrich
Anti-phospho-STAT1 (Ser 727) Antibody, clone 12C5, clone 12C5, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Interferon-γ antibody produced in mouse, clone 25718, purified immunoglobulin, lyophilized powder
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1
Sigma-Aldrich
Anti-Interferon-α/β Receptor Chain 2 Antibody, clone MMHAR-2, clone MMHAR-2, Chemicon®, from mouse