Merck
  • The white-rot fungus, Phanerochaete chrysosporium, under combinatorial stress produces variable oil profiles following analysis of secondary metabolites.
The white-rot fungus, Phanerochaete chrysosporium, under combinatorial stress produces variable oil profiles following analysis of secondary metabolites.
Journal of applied microbiology (2021-01-24)
R Whiteford, I Nurika, T Schiller, G Barker
ABSTRACT
The effects of combinatorial stress on lipid production in Phanerochaete chrysosporium remain understudied. This species of white-rot fungi was cultivated on solid-state media while under variable levels of known abiotic and biotic stressors to establish the effect upon fungal oil profiles. Environmental stressors induced upon the fungus included the following: temperature, nutrient limitation and interspecies competition to assess impact upon oil profiles. Fatty acid type and its concentration were determined using analytical methods of gas chromatography and mass spectrometry. Growth rate under stress was established using high-performance liquid chromatography with ergosterol as the biomarker. Fungi grown on solid-state agar were able to simultaneously produce short- and long-chain fatty acids which appeared to be influenced by nutritional composition as well as temperature. Addition of nitrogen supplements increased the growth rate, but lipid dynamics remained unchanged. Introducing competition-induced stress had significantly altered the production of certain fatty acids beyond that of the monoculture while under nutrient-limiting conditions. Linoleic acid concentrations, for example, increased from an average of 885 ng μl-1 at monoculture towards 13 820 ng μl-1 at co-culture, following 7 days of incubation. Interspecies competition produced the most notable impact on lipid production for solid-state media cultivated fungi while the addition of nitrogen supplementation presented growth and lipid accumulation to be uncorrelated. Combinatorial stress therefore influences the yield of overall lipid production as well as the number of intermediate fatty acids produced, deriving similar oil profiles to the composition of vegetable and fish oils. Fungal secondary metabolism remains highly sensitive following combinatorial stress. The outcome impacts the research towards optimizing fungal oil profiles for biomass and nutrition. Future investigations on fungal stress tolerance mechanisms need to address these environmental factors throughout the experimental design.
Sigma-Aldrich
Tripentadecanoin, ≥99%
Pricing and availability is not currently available.