Merck
  • Expression Pattern of iNOS, BCL-2 and MMP-9 in the Hip Synovium Tissue of Patients with Osteoarthritis.
Expression Pattern of iNOS, BCL-2 and MMP-9 in the Hip Synovium Tissue of Patients with Osteoarthritis.
International journal of molecular sciences (2021-02-06)
Davor Caric, Sandra Zekic Tomas, Natalija Filipovic, Violeta Soljic, Benjamin Benzon, Sandro Glumac, Ivan Rakovac, Katarina Vukojevic
ABSTRACT
Hip osteoarthritis (HOA) is characterized by degradation of the cartilage and synovitis. However, the pathohistological effects of synovial tissue inflammation on HOA are not clear. The aim of this study was to evaluate the expression of iNOS, BCL-2 and MMP-9 markers in different synovial cell populations. A total of 32 patients were evaluated retrospectively. Age, sex, height, weight, body mass index were recorded and lymphocyte, fibrocytes and macrophages were analysed in tissue sections. Osteoarthritis cartilage histopathology assessment system (OARSI), Western Ontario and McMaster Universities Osteoarthritis Index (WOMAC), Krenn score, Harris Hip Score (HHS) and Kellgren-Lawrence (K-L) grading of the hip joints were performed. Total hip arthroplasty was performed on 32 patients and controls. Patients were divided into two groups according to their disease severity. The tissues were immunohistochemically analysed. K-L grade and Krenn score differ between all three groups, but also between moderate and severe OA. Synovial lining cell layer, resident cells in stroma and especially inflammatory infiltration were increasing with severity of OA. iNOS expression in both intima and subintima was positively correlated with Krenn score in moderate and severe osteoarthritis (OA) groups. Expression of BCL-2 in intima of severe OA patients was positively correlated with Krenn score. In conclusion, iNOS, BCL-2 and MMP-9 are involved in the regulation of HOA. Our study indicates a relationship between the pathohistological features, the synovial inflammation and the cartilage condition at the time of hip replacement due to OA or femoral neck fracture.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, FITC conjugate, 2 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Chemicon®, from goat