Merck
  • Alteration of the serum N-glycome of mice locally exposed to high doses of ionizing radiation.
Alteration of the serum N-glycome of mice locally exposed to high doses of ionizing radiation.
Molecular & cellular proteomics : MCP (2012-11-14)
Thibault Chaze, Marie-Christine Slomianny, Fabien Milliat, Georges Tarlet, Tony Lefebvre-Darroman, Patrick Gourmelon, Eric Bey, Marc Benderitter, Jean-Claude Michalski, Olivier Guipaud
ABSTRACT
Exposure of the skin to ionizing radiation leads to characteristic reactions that will often turn into a pathophysiological process called the cutaneous radiation syndrome. The study of this disorder is crucial to finding diagnostic and prognostic bioindicators of local radiation exposure or radiation effects. It is known that irradiation alters the serum proteome content and potentially post-translationally modifies serum proteins. In this study, we investigated whether localized irradiation of the skin alters the serum glycome. Two-dimensional differential in-gel electrophoresis of serum proteins from a man and from mice exposed to ionizing radiation showed that potential post-translational modification changes occurred following irradiation. Using a large-scale quantitative mass-spectrometry-based glycomic approach, we performed a global analysis of glycan structures of serum proteins from non-irradiated and locally irradiated mice exposed to high doses of γ-rays (20, 40, and 80 Gy). Non-supervised descriptive statistical analyses (principal component analysis) using quantitative glycan structure data allowed us to discriminate between uninjured/slightly injured animals and animals that developed severe lesions. Decisional statistics showed that several glycan families were down-regulated whereas others increased, and that particular structures were statistically significantly changed in the serum of locally irradiated mice. The observed increases in multiantennary N-glycans and in outer branch fucosylation and sialylation were associated with the up-regulation of genes involved in glycosylation in the liver, which is the main producer of serum proteins, and with an increase in the key proinflammatory serum cytokines IL-1β, IL-6, and TNFα, which can regulate the expression of glycosylation genes. Our results suggest for the first time a role of serum protein glycosylation in response to irradiation. These protein-associated glycan structure changes might signal radiation exposure or effects.
Product Description
Supelco
ProteoMass Peptide MALDI-MS Calibration Kit, Peptide MALDI MS Calibration Standard