Merck
  • Remarkable transglycosylation activity of glycosynthase mutants of endo-D, an endo-β-N-acetylglucosaminidase from Streptococcus pneumoniae.
Remarkable transglycosylation activity of glycosynthase mutants of endo-D, an endo-β-N-acetylglucosaminidase from Streptococcus pneumoniae.
The Journal of biological chemistry (2012-02-10)
Shu-Quan Fan, Wei Huang, Lai-Xi Wang
ABSTRACT
Endo-β-N-acetylglucosaminidase from Streptococcus pneumoniae (Endo-D) is an endoglycosidase capable of hydrolyzing the Fc N-glycan of intact IgG antibodies after sequential removal of the sialic acid, galactose, and internal GlcNAc residues in the N-glycan. Endo-D also possesses transglycosylation activity with sugar oxazoline as the donor substrate, but the transglycosylation yield is low due to enzymatic hydrolysis of the donor substrate and the product. We report here our study on the hydrolytic and transglycosylation activity of recombinant Endo-D and its selected mutants. We found that Endo-D preferred core-fucosylated N-glycan for hydrolysis but favored nonfucosylated GlcNAc acceptor for transglycosylation. Several mutants showed significantly enhanced transglycosylation efficiency over the wild type enzyme. Two mutants (N322Q and N322A) were identified as typical glycosynthases that demonstrated remarkable transglycosylation activity with only marginal or no product hydrolysis activity. Kinetic studies revealed that the N322Q [corrected]and N322A glycosynthases had much higher catalytic efficiency for glycosylating the nonfucosylated GlcNAc acceptor. In comparison, the N322Q was much more efficient than N322A for transglycosylation. However, N322Q and N322A [corrected] could not take more complex N-glycan oxazoline as substrate for transglycosylation, indicating their strict substrate specificity. The usefulness of the N322Q glycosynthase was exemplified by its application for efficient glycosylation remodeling of IgG-Fc domain.
Sigma-Aldrich
Endoglycosidase F1 from Elizabethkingia miricola, recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F2 from Elizabethkingia miricola, recombinant, expressed in E. coli, 20 U/mg
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution
Supelco
ProteoMass Peptide MALDI-MS Calibration Kit, Peptide MALDI MS Calibration Standard
Sigma-Aldrich
Endoglycosidase F3 from Elizabethkingia miricola, recombinant, expressed in E. coli, 30 U/mg