Merck
  • Systematic mapping of protein-metabolite interactions in central metabolism of Escherichia coli.
Systematic mapping of protein-metabolite interactions in central metabolism of Escherichia coli.
Molecular systems biology (2019-08-30)
Maren Diether, Yaroslav Nikolaev, Frédéric Ht Allain, Uwe Sauer
ABSTRACT
Metabolite binding to proteins regulates nearly all cellular processes, but our knowledge of these interactions originates primarily from empirical in vitro studies. Here, we report the first systematic study of interactions between water-soluble proteins and polar metabolites in an entire biological subnetwork. To test the depth of our current knowledge, we chose to investigate the well-characterized Escherichia coli central metabolism. Using ligand-detected NMR, we assayed 29 enzymes towards binding events with 55 intracellular metabolites. Focusing on high-confidence interactions at a false-positive rate of 5%, we detected 98 interactions, among which purine nucleotides accounted for one-third, while 50% of all metabolites did not interact with any enzyme. In contrast, only five enzymes did not exhibit any metabolite binding and some interacted with up to 11 metabolites. About 40% of the interacting metabolites were predicted to be allosteric effectors based on low chemical similarity to their target's reactants. For five of the eight tested interactions, in vitro assays confirmed novel regulatory functions, including ATP and GTP inhibition of the first pentose phosphate pathway enzyme. With 76 new candidate regulatory interactions that have not been reported previously, we essentially doubled the number of known interactions, indicating that the presently available information about protein-metabolite interactions may only be the tip of the iceberg.
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
L-(−)-Malic acid sodium salt, 95-100% (enzymatic)
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Sigma-Aldrich
Acetyl coenzyme A trisodium salt, ≥93% (HPLC), powder
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Anti-EGFR Antibody, clone LA22, clone LA22, Upstate®, from mouse
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
D-(−)-3-Phosphoglyceric acid disodium salt, ≥93% dry basis (enzymatic), powder
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Flavin adenine dinucleotide disodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%