Merck
  • Novel positive allosteric modulators of A2B adenosine receptor acting as bone mineralisation promoters.
Novel positive allosteric modulators of A2B adenosine receptor acting as bone mineralisation promoters.
Journal of enzyme inhibition and medicinal chemistry (2020-12-19)
Elisabetta Barresi, Chiara Giacomelli, Laura Marchetti, Emma Baglini, Silvia Salerno, Giovanni Greco, Federico Da Settimo, Claudia Martini, Maria Letizia Trincavelli, Sabrina Taliani
ABSTRACT
Small-molecules acting as positive allosteric modulators (PAMs) of the A2B adenosine receptor (A2B AR) could potentially represent a novel therapeutic strategy for pathological conditions characterised by altered bone homeostasis, including osteoporosis. We investigated a library of compounds (4-13) exhibiting different degrees of chemical similarity with three indole derivatives (1-3), which have been recently identified by us as PAMs of the A2B AR able to promote mesenchymal stem cell differentiation and bone formation. Evaluation of mineralisation activity of 4-13 in the presence and in the absence of the agonist BAY60-6583 allowed the identification of lead compounds with therapeutic potential as anti-osteoporosis agents. Further biological characterisation of one of the most performing compounds, the benzofurane derivative 9, confirmed that such a molecule behaves as PAM of the A2B AR.
Sigma-Aldrich
Mesenchymal Stem Cell Osteogenic Differentiation Medium, Ready-to-use kit including Basal Medium and SupplementMix, 100 ml
Sigma-Aldrich
Human Mesenchymal Stem Cells (Bone Marrow), Human mesenchymal stem cells derived from bone marrow that have a capacity for expansion in vitro while maintaining their potential for differentiation to multiple lineages including adipocytes, osteoblasts and chondrocytes.