Merck
  • Triacylglycerol mobilization is suppressed by brefeldin A in Chlamydomonas reinhardtii.
Triacylglycerol mobilization is suppressed by brefeldin A in Chlamydomonas reinhardtii.
Plant & cell physiology (2013-07-23)
Naohiro Kato, Trung Dong, Michael Bailey, Tony Lum, Drury Ingram
ABSTRACT
Brefeldin A suppresses vesicle trafficking by inhibiting exchange of GDP for GTP in ADP-ribosylation factor. We report that brefeldin A suppresses mobilization of triacylglycerols in Chlamydomonas reinhardtii, a model organism of green microalgae. Analyses revealed that brefeldin A causes Chlamydomonas to form lipid droplets in which triacylglycerols accumulate in a dose-dependent manner. Pulse labeling experiment using fluorescent fatty acids suggested that brefeldin A inhibits the cells from degrading fatty acids. The experiment also revealed that the cells transiently form novel compartments that accumulate exogenously added fatty acids in the cytoplasm, designated fatty acid-induced microbodies (FAIMs). Brefeldin A up-regulates the formation of FAIMs, whereas nitrogen deprivation that up-regulates triacylglycerol synthesis in Chlamydomonas does not cause the cells to form FAIMs. These results underscore the role of the vesicle trafficking machinery in triacylglycerol metabolism in green microalgae.
Product Description
Supelco
Starch (HK) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.