Merck
  • Adipocyte Piezo1 mediates obesogenic adipogenesis through the FGF1/FGFR1 signaling pathway in mice.
Adipocyte Piezo1 mediates obesogenic adipogenesis through the FGF1/FGFR1 signaling pathway in mice.
Nature communications (2020-05-10)
ShengPeng Wang, Shuang Cao, Malika Arhatte, Dahui Li, Yue Shi, Sabrina Kurz, Jiong Hu, Lei Wang, Jingchen Shao, Ann Atzberger, Zheng Wang, Changhe Wang, Weijin Zang, Ingrid Fleming, Nina Wettschureck, Eric Honoré, Stefan Offermanns
ABSTRACT
White adipose tissue (WAT) expansion in obesity occurs through enlargement of preexisting adipocytes (hypertrophy) and through formation of new adipocytes (adipogenesis). Adipogenesis results in WAT hyperplasia, smaller adipocytes and a metabolically more favourable form of obesity. How obesogenic WAT hyperplasia is induced remains, however, poorly understood. Here, we show that the mechanosensitive cationic channel Piezo1 mediates diet-induced adipogenesis. Mice lacking Piezo1 in mature adipocytes demonstrated defective differentiation of preadipocyte into mature adipocytes when fed a high fat diet (HFD) resulting in larger adipocytes, increased WAT inflammation and reduced insulin sensitivity. Opening of Piezo1 in mature adipocytes causes the release of the adipogenic fibroblast growth factor 1 (FGF1), which induces adipocyte precursor differentiation through activation of the FGF-receptor-1. These data identify a central feed-back mechanism by which mature adipocytes control adipogenesis during the development of obesity and suggest Piezo1-mediated adipocyte mechano-signalling as a mechanism to modulate obesity and its metabolic consequences.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
PD 173074, ≥96% (HPLC), powder
Sigma-Aldrich
Human BMP2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Millipore
5-Bromo-2′-deoxyuridine, Thymidine analog. Useful for the study of DNA synthesis where it is incorporated into DNA in place of thymidine.