Merck
  • Arp2/3 and Mena/VASP Require Profilin 1 for Actin Network Assembly at the Leading Edge.
Arp2/3 and Mena/VASP Require Profilin 1 for Actin Network Assembly at the Leading Edge.
Current biology : CB (2020-05-30)
Kristen Skruber, Peyton V Warp, Rachael Shklyarov, James D Thomas, Maurice S Swanson, Jessica L Henty-Ridilla, Tracy-Ann Read, Eric A Vitriol
ABSTRACT
Cells have many types of actin structures, which must assemble from a common monomer pool. Yet, it remains poorly understood how monomers are distributed to and shared between different filament networks. Simplified model systems suggest that monomers are limited and heterogeneous, which alters actin network assembly through biased polymerization and internetwork competition. However, less is known about how monomers influence complex actin structures, where different networks competing for monomers overlap and are functionally interdependent. One example is the leading edge of migrating cells, which contains filament networks generated by multiple assembly factors. The leading edge dynamically switches between the formation of different actin structures, such as lamellipodia or filopodia, by altering the balance of these assembly factors' activities. Here, we sought to determine how the monomer-binding protein profilin 1 (PFN1) controls the assembly and organization of actin in mammalian cells. Actin polymerization in PFN1 knockout cells was severely disrupted, particularly at the leading edge, where both Arp2/3 and Mena/VASP-based filament assembly was inhibited. Further studies showed that in the absence of PFN1, Arp2/3 no longer localizes to the leading edge and Mena/VASP is non-functional. Additionally, we discovered that discrete stages of internetwork competition and collaboration between Arp2/3 and Mena/VASP networks exist at different PFN1 concentrations. Low levels of PFN1 caused filopodia to form exclusively at the leading edge, while higher concentrations inhibited filopodia and favored lamellipodia and pre-filopodia bundles. These results demonstrate that dramatic changes to actin architecture can be made simply by modifying PFN1 availability.
Sigma-Aldrich
Anti-Mena Antibody, clone A351F7D9, clone A351F7D9, from mouse
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Anti-beta-Actin Antibody, clone 4C2, clone 4C2, from mouse
Sigma-Aldrich
Anti-gamma-Actin/ACTG1 Antibody, clone 2A3, clone 2A3, from mouse
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder