Merck
  • Global Landscape and Dynamics of Parkin and USP30-Dependent Ubiquitylomes in iNeurons during Mitophagic Signaling.
Global Landscape and Dynamics of Parkin and USP30-Dependent Ubiquitylomes in iNeurons during Mitophagic Signaling.
Molecular cell (2020-03-07)
Alban Ordureau, Joao A Paulo, Jiuchun Zhang, Heeseon An, Kirby N Swatek, Joe R Cannon, Qiaoqiao Wan, David Komander, J Wade Harper
ABSTRACT
The ubiquitin ligase Parkin, protein kinase PINK1, USP30 deubiquitylase, and p97 segregase function together to regulate turnover of damaged mitochondria via mitophagy, but our mechanistic understanding in neurons is limited. Here, we combine induced neurons (iNeurons) derived from embryonic stem cells with quantitative proteomics to reveal the dynamics and specificity of Parkin-dependent ubiquitylation under endogenous expression conditions. Targets showing elevated ubiquitylation in USP30-/- iNeurons are concentrated in components of the mitochondrial translocon, and the ubiquitylation kinetics of the vast majority of Parkin targets are unaffected, correlating with a modest kinetic acceleration in accumulation of pS65-Ub and mitophagic flux upon mitochondrial depolarization without USP30. Basally, ubiquitylated translocon import substrates accumulate, suggesting a quality control function for USP30. p97 was dispensable for Parkin ligase activity in iNeurons. This work provides an unprecedented quantitative landscape of the Parkin-modified ubiquitylome in iNeurons and reveals the underlying specificity of central regulatory elements in the pathway.
Sigma-Aldrich
1-Butyl-3-methylimidazolium hexafluorophosphate, ≥97.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Chloroacetamide, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
EPPS, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Chloroacetamide, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-RHOT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
PR-619, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-March5 Antibody, serum, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)