Merck
  • Development of a novel model for comparative evaluation of intranasal pharmacokinetics and effects of anti-allergic nasal sprays.
Development of a novel model for comparative evaluation of intranasal pharmacokinetics and effects of anti-allergic nasal sprays.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2011-09-29)
Daniel Baumann, Claus Bachert, Petra Högger
ABSTRACT
For locally acting drugs, an extended residence time in the nasal cavity is desirable and related to a prolonged effect. We sought to develop a model for comparative determination of intranasal pharmacokinetics. We embedded human respiratory tissue into a solid matrix and coated the surface with artificial nasal fluid. Nasal spray suspensions of fluticasone propionate (FP) and budesonide (Bud) as well as a solution of azelastine hydrochloride (AZ) were applied onto the surface and removed after 30 min to simulate mucociliary clearance. As exemplary anti-inflammatory measure, we evaluated the inhibition of IL-8 release from epithelial cells. FP and Bud were initially bound to the same extent to the tissue gel while AZ displayed a more 4-fold higher binding than FP or Bud. After equilibrium with plasma, approximately 5-fold higher tissue concentrations of AZ compared to FP and 77-fold higher levels in relation to Bud were determined. This tissue retention revealed an excellent correlation with the volume of distribution of the respective drugs (r=0.9999, p ≤ 0.05). The inhibitory effect of FP on IL-8 release was approximately 5-fold more pronounced compared to AZ. The present model realistically mirrors conditions in vivo where solubility and tissue absorption of intranasally applied drugs compete with mucociliary clearance mechanisms.
Sigma-Aldrich
Azelastine hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.
Azelastine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.