Merck
  • Association of TIM-3 with BCLC Stage, Serum PD-L1 Detection, and Response to Transarterial Chemoembolization in Patients with Hepatocellular Carcinoma.
Association of TIM-3 with BCLC Stage, Serum PD-L1 Detection, and Response to Transarterial Chemoembolization in Patients with Hepatocellular Carcinoma.
Cancers (2020-01-19)
Maria Tampaki, Evangelos Ionas, Emilia Hadziyannis, Melanie Deutsch, Katerina Malagari, John Koskinas
ABSTRACT
Considering the increasing importance of immune checkpoints in tumor immunity we investigated the clinical relevance of serum T-cell immunoglobulin and mucin domain-3 (TIM-3) in patients with hepatocellular carcinoma (HCC). Serum TIM-3 levels were measured and their association with HCC stage and the detection of serum programmed death ligand-1 (PD-L1) were assessed. In patients submitted to transarterial chemoembolization (TACE), pre- and 1-week post-treatment TIM-3 levels were also evaluated. We studied 53 HCC patients with BCLC stages: 0 (5.7%), A (34%), B (32.1%), C (22.6%), and D (5.7%). The patients with advanced HCC (BCLC C) had significantly higher TIM-3 levels than patients with BCLC A (p = 0.009) and BCLC B (p = 0.019). TIM-3 levels were not associated with HCC etiology (p = 0.183). PD-L1 detection (9/53 patients) correlated with TIM-3 levels (univariate analysis, p = 0.047). In 33 patients who underwent TACE, post-treatment TIM-3 levels (231 pg/mL, 132-452) were significantly higher than pre-TACE levels (176 pg/mL, 110-379), (p = 0.036). Complete responders had higher post-TACE TIM-3 levels (534 pg/mL, 370-677) than partial responders (222 pg/mL, 131-368), (p = 0.028). Collectively, TIM-3 may have a role in anti-tumor immunity following TACE, setting a basis for combining immunotherapy and chemoembolization.
Sigma-Aldrich
Human TIM-3 ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
Human L1CAM ELISA Kit, for cell culture supernatants, plasma, and serum samples