Merck
  • Crystallographic and kinetic investigations of the covalent complex formed by a specific tetrapeptide aldehyde and the serine protease from Streptomyces griseus.
Crystallographic and kinetic investigations of the covalent complex formed by a specific tetrapeptide aldehyde and the serine protease from Streptomyces griseus.
Proceedings of the National Academy of Sciences of the United States of America (1979-01-01)
G D Brayer, L T Delbaere, M N James, C A Bauer, R C Thompson
PMID106392
ABSTRACT
X-ray crystallographic data show that a specific tetrapeptide aldehyde inhibitor (N-acetylprolylalanylprolylphenylalaninal) forms a stable, covalent, tetrahedral addition complex with the serine protease, SGPA, from Streptomyces griseus. Earlier proposals, based on kinetic measurements, for the covalent nature of such linkages are confirmed, and the difference electron density map of this aldehyde inhibitor indicates that a major conformational change of the histidyl-57 side chain occurs on inhibitor binding.
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder