Merck
  • KLF2 in Myeloid Lineage Cells Regulates the Innate Immune Response during Skeletal Muscle Injury and Regeneration.
KLF2 in Myeloid Lineage Cells Regulates the Innate Immune Response during Skeletal Muscle Injury and Regeneration.
iScience (2019-07-22)
Palanikumar Manoharan, Taejeong Song, Tatiana L Radzyukevich, Sakthivel Sadayappan, Jerry B Lingrel, Judith A Heiny
ABSTRACT
Skeletal muscle repair and regeneration after injury requires coordinated interactions between the innate immune system and the injured muscle. Myeloid cells predominate in these interactions. This study examined the role of KLF2, a zinc-finger transcription factor that regulates immune cell activation, in specifying myeloid cell functions during muscle regeneration. Loss of KLF2 in myeloid lineage cells (myeKlf2-/- mice) dramatically enhanced the initial inflammatory response to acute muscle injury (cardiotoxin). Injured muscles showed dramatically elevated expression of inflammatory mediators and greater numbers of infiltrating, pro-inflammatory monocytes that matured earlier into activated macrophages. Notably, the inflammatory phase resolved earlier and regeneration progressed to myogenesis, marked by elevated expression of factors that promote the formation of new fibers from satellite cells. Regeneration was completed earlier, with phenotypically normal adult fibers integrated into the muscle syncytium. These findings identify myeloid KLF2 as a key regulator of myeloid cell functions in adult skeletal muscle regeneration.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MYH3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution