Merck
  • Biophysical studies of the translation initiation pathway with immobilized mRNA analogs.
Biophysical studies of the translation initiation pathway with immobilized mRNA analogs.
Methods in enzymology (2007-10-05)
John E G McCarthy, Steven Marsden, Tobias von der Haar
ABSTRACT
A growing number of biophysical techniques use immobilized reactants for the quantitative study of macromolecular reactions. Examples of such approaches include surface plasmon resonance, atomic force microscopy, total reflection fluorescence microscopy, and others. Some of these methods have already been adapted for work with immobilized RNAs, thus making them available for the study of many reactions relevant to translation. Published examples include the study of kinetic parameters of protein/RNA interactions and the effect of helicases on RNA secondary structure. The common denominator of all of these techniques is the necessity to immobilize RNA molecules in a functional state on solid supports. In this chapter, we describe a number of approaches by which such immobilization can be achieved, followed by two specific examples for applications that use immobilized RNAs.
Sigma-Aldrich
ExtrAvidin®, essentially salt-free, lyophilized powder
Pricing and availability is not currently available.
Roche
Streptavidin, recombinant, liquid, ≥17 units/mg protein, pH range 5.0-9.0
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Pricing and availability is not currently available.