Merck
  • PLD1 regulates adipogenic differentiation through mTOR - IRS-1 phosphorylation at serine 636/639.
PLD1 regulates adipogenic differentiation through mTOR - IRS-1 phosphorylation at serine 636/639.
Scientific reports (2016-11-23)
Hae-In Song, Mee-Sup Yoon
ABSTRACT
Phospholipase D1 (PLD1) plays a known role in several differentiation processes, but its role in adipogenic differentiation remains unknown. In the present study, we identified PLD1 as a negative regulator of adipogenic differentiation. We showed that PLD activity was downregulated by both 3-Isobutyl-1-methylxanthine (IBMX) and insulin upon induction of differentiation in 3T3-L1 adipogenic cells. In line with this observation, PLD activity decreased in both high fat diet (HFD)-fed mice and ob/ob mice. We also found that differentiation of 3T3-L1 preadipocytes was enhanced by the depletion of PLD1 levels or inhibition of PLD1 activity by VU0155069, a PLD1-specific inhibitor. Conversely, treatment with phosphatidic acid (PA), a PLD product, and overexpression of PLD1 both caused a decrease in adipogenic differentiation. Moreover, the elevated differentiation in PLD1-knockdown 3T3-L1 cells was reduced by either PA treatment or PLD1 expression, confirming negative roles of PLD1 and PA in adipogenic differentiation. Further investigation revealed that PA displaces DEP domain-containing mTOR-interacting protein (DEPTOR) from mTORC1, which subsequently phosphorylates insulin receptor substrate-1 (IRS-1) at serine 636/639 in 3T3-L1 cells. Taken together, our findings provide convincing evidence for a direct role of PLD1 in adipogenic differentiation by regulating IRS-1 phosphorylation at serine 636/639 through DEPTOR displacement and mTOR activation.
Sigma-Aldrich
VU0155069, ≥98% (HPLC)