Merck
  • Glycoproteins in circulating immune complexes are biomarkers of patients with Indian PKDL: A study from endemic districts of West Bengal, India.
Glycoproteins in circulating immune complexes are biomarkers of patients with Indian PKDL: A study from endemic districts of West Bengal, India.
PloS one (2018-02-09)
Priyank Jaiswal, Souvik Datta, Bikash Sardar, Surya Jyoti Chaudhuri, Dipankar Maji, Manab Ghosh, Bibhuti Saha, Sumi Mukhopadhyay
ABSTRACT
Post Kala Azar Dermal Leishmaniasis (PKDL) occurs as dermal consequence of previous Visceral Leishmaniasis (VL) infection and serves as an important reservoir for transmission of VL. Diagnosis of PKDL is often challenging for its symptomatic resemblance to other co-endemic diseases like Leprosy or Vitiligo. Parasitological examination by slit-skin smear and culture are the standard methods but lack high sensitivity. Thus, for efficient control of VL, reliable diagnostic and prognostic assay of PKDL are required. Previously, glycoproteins (9-OAcSA) have been reported as promising biomarkers of Indian VL patients. However, till date, the status of glycans in Indian PKDL patients remains unexplored. Accordingly, in this study, the glyco-profile of PKDL Circulating Immune Complexes (CICs) as compared to other cross diseases like Vitiligo and Leprosyhas been investigated. Further, a novel Glyco CIC assay has been developed for efficient Indian PKDL patient diagnosis. In the present study, 90 PKDL patients were enrolled from 3 VL endemic districts of West Bengal during 2015-16. Glycosylation profile of isolated CICs from sera of PKDL patients were initially analyzed through gradient SDS gel electrophoresis followed by PAS silver double staining, which revealed the presence of several glycan rich PKDL specific proteins of varying molecular weights. To further characterize the glyco-profile of acid dissociated affinity purified immuno-reactive antigens present in the CICs, glycosylation was demonstrated in these purified CIC antigens by DIG glycan differentiation kit with or without glycosidase as well as neuraminidase treatment. Diagnostic evaluation of the newly developed colorimetric Glyco CIC assay through Receiver Operating Characteristic (ROC) curve analysis revealed excellent (0.99) AUC value as compared to other conventional serodiagnostic assays like PEG CIC, Parasite ELISA (IgG and IgM). Additionally, longitudinal monitoring of 18 PKDL patients further revealed its good prognostic utility. These results highlight the glycosylation status of CICs among Indian PKDL patients present in all the studied endemic districts of West Bengal. These PKDL biomarkers were completely absent in cross diseases like Vitiligo and Leprosy. Further, the newly developed Glyco CIC assay had an improved sensitivity of 95.6%, specificity of 99.3%, NPV of 97.1% and PPV of 98.9%.
Sigma-Aldrich
Anti-Human IgM (μ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody