Skip to Content
Merck
Zopiclone British Pharmacopoeia (BP) Reference Standard

Zopiclone

Zopiclone British Pharmacopoeia (BP) Reference Standard
Synonym(s):
4-Methyl-1-piperazinecarboxylic acid 6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, Imovane
Empirical Formula (Hill Notation):
C17H17ClN6O3
CAS Number:
Molecular Weight:
388.81
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing