
Zirconium(IV) propoxide solution

Synonym(s):
Tetrapropyl zirconate
Linear Formula:
Zr(OCH2CH2CH3)4
CAS Number:
Molecular Weight:
327.57
Products
Boiling Point (°C)
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing