Skip to Content
Merck
Vanadium rod, 11 mm diameter, length 100 mm, purity 99.8%

Vanadium

Vanadium rod, 11 mm diameter, length 100 mm, purity 99.8%
Synonym(s):
Vanadium
Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Element
Quality Segment
Showing 1-20 of 187
Page 1 of 10
Compare
Product Number
Description
Pricing
Page 1 of 10