
Valsartan Related Compound A

Synonym(s):
D-Valsartan, (R)-N-Valeryl-N-([2′-(1H-tetrazole-5-yl)-biphenyl-4-yl]methyl)valine, N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-D-valine
Empirical Formula (Hill Notation):
C24H29N5O3
Molecular Weight:
435.52
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2