
Triton™ X-100 solution

Synonym(s):
Triton™ X-100, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether
Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Products
Product Category
Brand
pH Value
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing