
Trichlorfon

Synonym(s):
(2,2,2-Trichloro-1-hydroxyethyl)phosphonic acid dimethyl ester, Metrifonate
Empirical Formula (Hill Notation):
C4H8Cl3O4P
CAS Number:
Molecular Weight:
257.44
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade