
Tricaprylin

Synonym(s):
Glyceryl trioctanoate, 1,2,3-Tricapryloylglycerol, 1,2,3-Trioctanoylglycerol, Glycerol tricaprylate, Glycerol trioctanoate, Glyceryl tricaprylate, Tricaprylin, Trioctanoin
Linear Formula:
[CH3(CH2)6COOCH2]2CHOCO(CH2)6CH3
CAS Number:
Molecular Weight:
470.68
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing