
Thimerosal Related Compound A

Synonym(s):
2,2′-Dithiodibenzoic acid, 2-Carboxyphenyl disulfide, Bis(2-carboxyphenyl) disulfide, Dithiosalicylic acid
Linear Formula:
S2(C6H4CO2H)2
CAS Number:
Molecular Weight:
306.36
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2