
Tenax®TA Thermal Desorption Tube

Synonym(s):
Tenax® TA
Products
Brand
Sustainability
Feature
Agency
ID (mm)
Length (in)
OD (mm)
Compatibility
Quality Segment
Application
Package Size
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing