
Tacrolimus Related Compound A

Synonym(s):
(E)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-Hexadecahydro-5,19-dihydroxy-3-[(E)-2-(4-hydroxy-3-methoxycyclohexyl)-1-methylvinyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21-(4H,23H)-tetrone
Empirical Formula (Hill Notation):
C43H69NO12
CAS Number:
Molecular Weight:
792.01
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing