![Sodium phosphate dibasic BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri™](/deepweb/assets/sigmaaldrich/product/structures/140/340/849261c2-36b2-4cdf-8515-ef928b197903/640/849261c2-36b2-4cdf-8515-ef928b197903.png)
Sodium phosphate dibasic
![Sodium phosphate dibasic BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri™](/deepweb/assets/sigmaaldrich/product/structures/140/340/849261c2-36b2-4cdf-8515-ef928b197903/640/849261c2-36b2-4cdf-8515-ef928b197903.png)
Synonym(s):
sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate, Sodium phosphate dibasic
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2