
Sodium 1-propanesulfonate monohydrate

Synonym(s):
1-Propanesulfonic acid sodium salt monohydrate
Linear Formula:
CH3CH2CH2SO3Na · H2O
CAS Number:
Molecular Weight:
164.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2