
Silver sulfadiazine

Synonym(s):
Silver(I) sulfadiazine, Sulfadiazine silver
Empirical Formula (Hill Notation):
C10H9AgN4O2S
CAS Number:
Molecular Weight:
357.14
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2