Skip to Content
Merck
Silica gel high-purity grade (7754), pore size 60 Å, 70-230 mesh

Silica gel Inorganic Sorbent

Silica gel high-purity grade (7754), pore size 60 Å, 70-230 mesh
Synonym(s):
Silica gel
CAS Number:
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Purity
Separation Technique
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-20 of 40
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2