
(S)-3-Pyrrolidinol

Synonym(s):
(S)-3-Hydroxypyrrolidine
Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
Products
Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing