
Roxarsone

Synonym(s):
4-Hydroxy-3-nitrobenzenearsonic acid, 2-Nitrophenol-4-arsonic acid
Linear Formula:
HOC6H3(NO2)AsO3H2
CAS Number:
Molecular Weight:
263.04
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing