Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator
Products
Product Category
rotors (14)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
EP022822228
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-35-6-30, (6 x 15 mL conical tube and 50 mL conical tubes )
Expand
View Pricing
EP022822047
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-70-11, non aerosol tight, to hold, 70 x 1.5/2.0 mL reaction vessels
Expand
View Pricing
EP022822080
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-72-8, non aerosol tight, to hold, 72 x 0.5 mL reaction vessels
Expand
View Pricing
RF454811-EP
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-48-11, non aerosol tight, to hold, 48 x 1.5/2.0 mL reaction vessels
Expand
View Pricing
EP022822225
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-8-17, non aerosol tight, (8 x 15 mL conical tubes)
Expand
View Pricing
EP022822187
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-35-8-24, non aerosol tight, (8 x 25.0 mL flat-bottom tubes)
Expand
View Pricing
EP022822209
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-40-36-12, non aerosol tight, (36 x 1.5 mL flat-bottom tubes)
Expand
View Pricing
RA2VC-EP
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
swing-out rotor, A-2-VCm non aerosol tight
Expand
View Pricing
EP022822128
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-36-15, non aerosol tight, to hold, 36 x 3.0/5.0 mL flat-bottom tubes
Expand
View Pricing
EP022822179
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-50-8-18, non aerosol tight, (8 x 10 mL - 18 mL round-bottom tubes (18 x 105-128 mm))
Expand
View Pricing
EP022822136
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-16-20, non aerosol tight, (16 x 6.5/10.0 mL flat-bottom tubes)
Expand
View Pricing
EP022822144
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-45-24-12, non aerosol tight, (24 x 6.0/8.0 mL round-bottom tubes)
Expand
View Pricing
EP022822233
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-50-8-16, non aerosol tight, (8 x 8- 12 mL round-bottom tubes)
Expand
View Pricing
EP022822161
Rotor for Eppendorf
®
Vacufuge
®
Plus Concentrator,
F-40-18-19, non aerosol tight, (8 x 10.0 mL flat-bottom tubes)
Expand
View Pricing