
Regorafenib monohydrate

Synonym(s):
4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-3-fluorophenoxy]-N-methyl-2-pyridinecarboxamide monohydrate
Empirical Formula (Hill Notation):
C21H15ClF4N4O3 · H2O
CAS Number:
Molecular Weight:
500.83
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing