
Rebaudioside C

Synonym(s):
(4α)-13-[(O-6-Deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid β-D-glucopyranosyl ester, Dulcoside B
Empirical Formula (Hill Notation):
C44H70O22
CAS Number:
Molecular Weight:
951.01
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing