Skip to Content
Merck
(R)-3-Pyrrolidinol hydrochloride 98%

(R)-3-Pyrrolidinol hydrochloride

(R)-3-Pyrrolidinol hydrochloride 98%
Synonym(s):
(R)-3-Hydroxypyrrolidine hydrochloride
Empirical Formula (Hill Notation):
C4H9NO · HCl
CAS Number:
Molecular Weight:
123.58

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing