Skip to Content
Merck
(R)-(−)-1-Indanol 99%

(R)-(−)-1-Indanol

(R)-(−)-1-Indanol 99%
Synonym(s):
R(−)-1-Hydroxyindan
Empirical Formula (Hill Notation):
C9H10O
CAS Number:
Molecular Weight:
134.18

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing