Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
VISTA
™
test tube
Synonym(s):
Corning
®
70800
Products
Product Category
tubes (8)
Color
Manufacturer
Diameter (mm)
Volume (ml)
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS7080016
Pyrex
®
VISTA
™
test tube,
diam. × L 16 mm × 150 mm
Expand
View Pricing
CLS7080013
Pyrex
®
VISTA
™
test tube,
diam. × L 13 mm × 100 mm
Expand
View Pricing
CLS7080012
Pyrex
®
VISTA
™
test tube,
diam. × L 12 mm × 75 mm
Expand
View Pricing
CLS7080020
Pyrex
®
VISTA
™
test tube,
diam. × L 20 mm × 150 mm
Expand
View Pricing
CLS7080018
Pyrex
®
VISTA
™
test tube,
diam. × L 18 mm × 150 mm
Expand
View Pricing
CLS7080025
Pyrex
®
VISTA
™
test tube,
diam. × L 25 mm × 150 mm
Expand
View Pricing
CLS7080015
Pyrex
®
VISTA
™
test tube,
diam. × L 15 mm × 125 mm
Expand
View Pricing
CLS7080025X
Pyrex
®
VISTA
™
test tube,
diam. × L 25 mm × 200 mm
Expand
View Pricing