Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
snap cap
Synonym(s):
Corning
®
7666
Products
Product Category
caps (9)
Color
Manufacturer
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
CLS7666100
Pyrex
®
snap cap,
snap cap size 100
Expand
View Pricing
CLS766650
Pyrex
®
snap cap,
snap cap size 50
Expand
View Pricing
CLS7666500
Pyrex
®
snap cap,
snap cap size 500
Expand
View Pricing
CLS76661L
Pyrex
®
snap cap,
snap cap size 1,000
Expand
View Pricing
CLS766625
Pyrex
®
snap cap,
snap cap size 25
Expand
View Pricing
CLS766610
Pyrex
®
snap cap,
snap cap size 10
Expand
View Pricing
CLS76662L
Pyrex
®
snap cap,
snap cap size 2,000
Expand
View Pricing
CLS7666200
Pyrex
®
snap cap,
snap cap size 200
Expand
View Pricing
CLS7666250
Pyrex
®
snap cap,
snap cap size 250
Expand
View Pricing