Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
powder funnel
Synonym(s):
Corning
®
6220
Products
Product Category
funnels (5)
Manufacturer
ID (mm)
Length (mm)
OD (mm)
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS6220100
Pyrex
®
powder funnel,
Expand
View Pricing
CLS622065
Pyrex
®
powder funnel,
Expand
View Pricing
CLS622075
Pyrex
®
powder funnel,
Expand
View Pricing
CLS6220125
Pyrex
®
powder funnel,
Expand
View Pricing
CLS6220150
Pyrex
®
powder funnel,
Expand
View Pricing