Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Allihn condenser, with male and female joints
Products
Product Category
desiccators (1)
Manufacturer
Length (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS2480500
Pyrex
®
Allihn condenser, with male and female joints,
jacket L 500 mm, joint: ST/NS 24/40
Expand
View Pricing
CLS2480300
Pyrex
®
Allihn condenser, with male and female joints,
jacket L 300 mm, joint: ST/NS 24/40
Expand
View Pricing
CLS2480400
Pyrex
®
Allihn condenser, with male and female joints,
jacket L 400 mm, joint: ST/NS 24/40
Expand
View Pricing
CLS2480600
Pyrex
®
Allihn condenser, with male and female joints,
jacket L 600 mm, joint: ST/NS 29/42
Expand
View Pricing
CLS2480200
Pyrex
®
Allihn condenser, with male and female joints,
jacket L 200 mm, joint: ST/NS 19/38
Expand
View Pricing